Abnova
Manufacturer Code:MAB11202
Catalog # 89-109-371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AT15E5 |
Immunogen | Recombinant protein corresponding to amino acids 1-475 of human CNDP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage,aliquot and store at -20°C. Avoid repeated freezing and thawing cycles. |
Mouse monoclonal antibody raised against partial recombinant CNDP2.
For Research Use Only
Protein Aliases: Carnosine dipeptidase II; CN2; CNDP dipeptidase 2; CPGL; Cytosolic non-specific dipeptidase; cytosolic nonspecific dipeptidase; Epididymis secretory protein Li 13; FLJ10830; Glutamate carboxypeptidase-like protein 1; HsT2298; PEPA; Peptidase A
Gene Aliases: CN2; CNDP2; CPGL; HEL-S-13; HsT2298; PEPA
UniProt ID: (Human) Q96KP4
Entrez Gene ID: (Human) 55748
Molecular Function: deacetylase hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.