Invitrogen
Manufacturer Code:PA581361
Catalog # PIPA581361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse CNDP2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CN2 CPGL HsT2298 PEPA cytosolic non-specific dipeptidase cytosolic nonspecific dipeptidase glutamate carboxypeptidase-like protein 1 peptidase A; CNDP dipeptidase 2; Cytosolic non-specific dipeptidase; cytosolic nonspecific dipeptidase; Glutamate carboxypeptidase-like protein 1; peptidase 1; peptidase A
Gene Aliases: 0610010E05Rik; C76600; Cn2; Cndp2; Dip-2; Pep-1; Pep1
UniProt ID: (Mouse) Q9D1A2
Entrez Gene ID: (Mouse) 66054
Molecular Function:
deacetylase
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.