Invitrogen
Manufacturer Code:PA544714
Catalog # PIPA544714
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CNDP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carnosine dipeptidase II; CNDP dipeptidase 2; CNDP dipeptidase 2 (metallopeptidase M20 family); Cytosolic non-specific dipeptidase; cytosolic nonspecific dipeptidase; cytosolic nonspecific dipeptidase 2; Epididymis secretory protein Li 13; Glutamate carboxypeptidase-like protein 1; peptidase 1; Peptidase A
Gene Aliases: 0610010E05Rik; C76600; CN2; CNDP2; CPGL; Dip-2; HEL-S-13; HsT2298; Pep-1; Pep1; PEPA; wu:fd20d11; wu:fe17f09; zgc:92569
UniProt ID: (Human) Q96KP4, (Rat) Q6Q0N1, (Mouse) Q9D1A2
Entrez Gene ID: (Rabbit) 100347984, (Horse) 100061697, (Human) 55748, (Dog) 476166, (Guinea pig) 100735444, (Rat) 291394, (Mouse) 66054, (Zebrafish) 327288
Molecular Function:
deacetylase
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.