Invitrogen
Manufacturer Code:MA529167
Catalog # PIMA529167
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombit Human CNDP1 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-Ala-His dipeptidase; carnosinase 1; Carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CN1 CPGL2 HsT2308 CNDP dipeptidase 1 beta-Ala-His dipeptidase carnosinase 1 glutamate carboxypeptidase-like protein 2 serum carnosinase; CNDP dipeptidase 1; Glutamate carboxypeptidase-like protein 2; Serum carnosinase
Gene Aliases: CN1; CNDP1; CPGL2; HsT2308; UNQ1915/PRO4380
UniProt ID: (Human) Q96KN2
Entrez Gene ID: (Human) 84735
Molecular Function:
deacetylase
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.