Proteintech
Manufacturer Code:177591AP150UL
Catalog # 50-173-1771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CNDP1 Fusion Protein Ag12127 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta Ala His dipeptidase Carnosine dipeptidase 1 CN1 CNDP dipeptidase 1 CNDP1 CPGL2 HsT2308 Serum carnosinase; Beta-Ala-His dipeptidase; carnosinase 1; Carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CNDP dipeptidase 1; Glutamate carboxypeptidase-like protein 2; Serum carnosinase
Gene Aliases: AI746433; CN1; CNDP1; CPGL2; HsT2308; UNQ1915/PRO4380
UniProt ID: (Human) Q96KN2, (Mouse) Q8BUG2, (Rat) Q66HG3
Entrez Gene ID: (Human) 84735, (Mouse) 338403, (Rat) 307212
Molecular Function: deacetylase hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.