Abnova
Manufacturer Code:H00112616M01A
Catalog # 89-107-529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S1 |
Immunogen | CMTM7 (AAH10116.11 a.a. &bsim 175 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant CMTM7.
For Research Use Only
Protein Aliases: chemokine-like factor super family 7; chemokine-like factor super family member 7 variant 2; chemokine-like factor superfamily 7; Chemokine-like factor superfamily member 7; CKLF-like MARVEL transmembrane domain containing 7; CKLF-like MARVEL transmembrane domain-containing protein 7; CKLFSF7; FLJ30992
Gene Aliases: CKLFSF7; CMTM7
UniProt ID: (Human) Q96FZ5
Entrez Gene ID: (Human) 112616
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.