Invitrogen
Manufacturer Code:PA568148
Catalog # PIPA568148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human CMTM7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chemokine-like factor super family 7; chemokine-like factor super family member 7 variant 2; chemokine-like factor superfamily 7; Chemokine-like factor superfamily member 7; CKLF-like MARVEL transmembrane domain containing 7; CKLF-like MARVEL transmembrane domain-containing protein 7; CKLFSF7 CKLF-like MARVEL transmembrane domain-containing protein 7 chemokine-like factor super family 7 chemokine-like factor super family member 7 variant 2 chemokine-like factor superfamily 7 chemokine-like factor superfamily member 7; LNV
Gene Aliases: AI481279; CKLFSF7; CMTM7; LNV
UniProt ID: (Human) Q96FZ5, (Mouse) Q9ESD6
Entrez Gene ID: (Human) 112616, (Mouse) 102545
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.