Novus Biologicals
Manufacturer Code:NBP257873
Catalog # NBP257873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ASIWLSYKISCVTQSTDAAV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chemokine-like factor super family 7; chemokine-like factor super family 7 chemokine-like factor super family member 7 variant 2 chemokine-like factor superfamily 7 Chemokine-like factor superfamily member 7 CKLF-like MARVEL transmembrane domain containing 7 CKLF-like MARVEL transmembrane domain-containing protein 7 CKLFSF7 FLJ30992; chemokine-like factor super family member 7 variant 2; chemokine-like factor superfamily 7; Chemokine-like factor superfamily member 7; CKLF-like MARVEL transmembrane domain containing 7; CKLF-like MARVEL transmembrane domain-containing protein 7
Gene Aliases: CKLFSF7; CMTM7
UniProt ID: (Human) Q96FZ5
Entrez Gene ID: (Human) 112616
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.