Invitrogen
Manufacturer Code:PA525130
Catalog # PIPA525130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 142-170 amino acids from the central region of human CMPK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial; cytidylate kinase 2; mitochondrial UMP-CMP kinase; Nucleoside-diphosphate kinase; thymidine monophosphate kinase 2; thymidylate kinase family LPS-inducible member; TMPK2 TYKi UMP-CMPK2 UMP-CMP kinase 2 mitochondrial cytidylate kinase 2 mitochondrial UMP-CMP kinase thymidine monophosphate kinase 2 thymidylate kinase family LPS-inducible member; UMP-CMP kinase 2, mitochondrial; UMP/CMP kinase
Gene Aliases: CMPK2; NDK; TMPK2; TYKi; UMP-CMPK2
UniProt ID: (Human) Q5EBM0
Entrez Gene ID: (Human) 129607
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.