Invitrogen
Manufacturer Code:MA526119
Catalog # PIMA526119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A2 |
Immunogen | Full length human recombit protein of CMPK1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CK; CMPK1; CMPK1 CMK CMPK UCK UMK UMPK UMP-CMP kinase Cytidine monophosphate kinase Cytidylate kinase Deoxycytidylate kinase Uridine monophosphate kinase Uridine monophosphate/cytidine monophosphate kinase UMP/CMP kinase UMP/CMPK; cytidine monophosphate (UMP-CMP) kinase 1, cytosolic; cytidine monophosphate kinase; cytidylate kinase; cytidylate kinase 1; dCMP kinase; Deoxycytidylate kinase; likely ortholog of H. sapiens UMP-CMP kinase (UMP-CMPK); Nucleoside-diphosphate kinase; UMP-CMP kinase; UMP/CMP kinase; UMP/CMPK; uridine monophosphate kinase; Uridine monophosphate/cytidine monophosphate kinase
Gene Aliases: 0610011D08Rik; CK; CMK; CMPK; CMPK1; UCK; UMK; UMP-CMPK; UMPK
UniProt ID: (Human) P30085, (Mouse) Q9DBP5
Entrez Gene ID: (Rhesus monkey) 710196, (Human) 51727, (Dog) 610291, (Mouse) 66588
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.