Novus Biologicals
Manufacturer Code:NBP271947IR
Catalog # NBP271947IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A1 |
Immunogen | Full length recombinant protein of human CMP kinase (NP_057392) produced in HEK293T cells. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CK; CMKEC 2.7.4.14 CMPKUMKUMP-CMP kinase cytidine monophosphate (UMP-CMP) kinase 1 cytosolic Cytidine monophosphate kinase Cytidylate kinaseUMP/CMP kinase Deoxycytidylate kinase EC 2.7.4 UMP/CMPK UMP-CMPK UMPKUCK Uridine monophosphate kinase Uridine monophosphate/cytidine monophosphate kinase; CMPK1; cytidine monophosphate (UMP-CMP) kinase 1, cytosolic; cytidylate kinase; dCMP kinase; Deoxycytidylate kinase; Nucleoside-diphosphate kinase; UMP-CMP kinase; UMP/CMP kinase; uridine monophosphate kinase; Uridine monophosphate/cytidine monophosphate kinase
Gene Aliases: CK; CMK; CMPK; CMPK1; UCK; UMK; UMP-CMPK; UMPK
UniProt ID: (Human) P30085
Entrez Gene ID: (Human) 51727
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.