Novus Biologicals
Manufacturer Code:NBP160012
Catalog # NBP160012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to NAT8B(N-acetyltransferase 8B (gene/pseudogene)) The peptide sequence was selected from the middle region of NAT8B. Peptide sequence SCFWVGESEEKVVGTVGALPVDDPTLREKRLQLFHLSVDNEHRGQGIAKA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetyltransferase 1; ATase1; Camello-like protein 2; Camello-like protein 2 CML2 EC 2.3.1 EC 2.3.1.- Hcml2 MGC97061 N-acetyltransferase 8B (GCN5-related putative gene/pseudogene) N-acetyltransferase 8B (gene/pseudogene) N-acetyltransferase Camello 2 NAT8BP probable N-acetyltransferase 8B; N-acetyltransferase 8B (GCN5-related, putative, gene/pseudogene); N-acetyltransferase Camello 2; probable N-acetyltransferase 8B; Putative N-acetyltransferase 8B
Gene Aliases: CML2; Hcml2; NAT8B; NAT8BP
UniProt ID: (Human) Q9UHF3
Entrez Gene ID: (Human) 51471
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.