Invitrogen
Manufacturer Code:PA599441
Catalog # PIPA599441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CMKLR1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chemerin chemokine-like receptor 1 chemerin receptor CHEMERINR chemokine receptor-like 1 chemokine-like receptor 1 CHEMR23 CMKLR1 Dez Gpcr27 G-protein coupled chemoattractant-like receptor G-protein coupled receptor 27 G-protein coupled receptor ChemR23 G-protein coupled receptor DEZ mcmklr1 orphan G-protein coupled receptor Dez resolvin E1 receptor RVER1; chemerin receptor; chemokine receptor-like 1; Chemokine-like receptor 1; G-protein coupled chemoattractant-like receptor; G-protein coupled receptor 27; G-protein coupled receptor ChemR23; G-protein coupled receptor DEZ; orphan G-protein coupled receptor, Dez; resolvin E1 receptor
Gene Aliases: CHEMERINR; CHEMR23; CMKLR1; DEZ; Gpcr27; mcmklr1; RVER1
UniProt ID: (Human) Q99788, (Mouse) P97468
Entrez Gene ID: (Human) 1240, (Mouse) 14747, (Rat) 60669
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.