Invitrogen
Manufacturer Code:PA5112976
Catalog # PIPA5112976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Clavesin-2 protein (1-327AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A330019N05Rik bA160A10.4 C6orf212 C6orf213 clathrin vesicle-associated Sec14 protein 2 clavesin 2 Clavesin-2 clavesin-2 {ECO:0000303 Clvs2 clvs2 {ECO:0000303 PubMed:19651769} retinaldehyde binding protein 1-like 2 Retinaldehyde-binding protein 1-like 2 retinaldehyde-binding protein 1-like protein 2 RGD1306801 RLBP1L2 si:ch211-199i15.4; clathrin vesicle-associated Sec14 protein 2; Clavesin-2; retinaldehyde binding protein 1-like 2; Retinaldehyde-binding protein 1-like 2; retinaldehyde-binding protein 1-like protein 2
Gene Aliases: A330019N05Rik; bA160A10.4; C6orf212; C6orf213; CLVS2; RLBP1L2
UniProt ID: (Human) Q5SYC1, (Mouse) Q8BG92
Entrez Gene ID: (Human) 134829, (Mouse) 215890
Molecular Function: dehydrogenase oxidoreductase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.