Novus Biologicals
Manufacturer Code:NBP297429AF700
Catalog # NB043348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human CLVS2 fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: clathrin vesicle-associated Sec14 protein 2; Clavesin-2; retinaldehyde binding protein 1-like 2; Retinaldehyde-binding protein 1-like 2
Gene Aliases: bA160A10.4; C6orf212; C6orf213; CLVS2; RLBP1L2
UniProt ID: (Human) Q5SYC1
Entrez Gene ID: (Human) 134829
Molecular Function: dehydrogenase oxidoreductase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.