OriGene
Manufacturer Code:CF808665
Catalog # 50-166-9005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 23-227 of human CLU produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AAG4 APO-J APOJ CLI CLU1 CLU2 KUB1 NA1/NA2 SGP-2 SGP2 SP-40 TRPM-2 TRPM2; Aging-associated gene 4 protein; aging-associated protein 4; Apo-J; ApoJalpha; ApoJbeta; Apolipoprotein J; CLI; Clusterin; Clusterin alpha chain; Clusterin beta chain; Complement cytolysis inhibitor; Complement cytolysis inhibitor a chain; Complement cytolysis inhibitor b chain; complement lysis inhibitor; Complement-associated protein SP-40,40; Ku70-binding protein 1; NA1/NA2; SGP-2; Sulfated glycoprotein 2; Testosterone-repressed prostate message 2; TRPM-2
Gene Aliases: AAG4; APO-J; APOJ; CLI; CLU; CLU1; CLU2; KUB1; NA1/NA2; SGP-2; SGP2; SP-40; TRPM-2; TRPM2
UniProt ID: (Human) P10909
Entrez Gene ID: (Human) 1191
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.