Proteintech
Manufacturer Code:664871IG150UL
Catalog # 50-173-6910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B3D7 |
Immunogen | CLTC Fusion Protein Ag25040 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CHC CHC17 Clathrin heavy chain 1 clathrin heavy chain (Hc) CLH 17 CLH17 CLTC CLTCL2 Hc KIAA0034; Clathrin heavy chain 1; Clathrin heavy chain on chromosome 17; clathrin, heavy chain (Hc); clathrin, heavy polypeptide (Hc); clathrin, heavy polypeptide-like 2; CLH-17
Gene Aliases: 3110065L21Rik; CHC; CHC17; CLH-17; CLH17; CLTC; CLTCL2; Hc; KIAA0034; R74732
UniProt ID: (Human) Q00610, (Rat) P11442, (Mouse) Q68FD5
Entrez Gene ID: (Human) 1213, (Pig) 100271929, (Rat) 54241, (Mouse) 67300
Molecular Function:
membrane traffic protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.