Proteintech
Manufacturer Code:662701IG150UL
Catalog # 50-173-6703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F9C2 |
Immunogen | CLTB Fusion Protein Ag24347 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Clathrin light chain B; Clathrin light chain B clathrin light chain (Lcb) CLTB LCB; clathrin light chain B pseudogene; clathrin, light chain (Lcb); clathrin, light chain B; clathrin, light polypeptide (Lcb); Lcb
Gene Aliases: CLTB; LCB
UniProt ID: (Human) P09497
Entrez Gene ID: (Human) 1212, (Pig) 100525860
Molecular Function: membrane traffic protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.