Invitrogen
Manufacturer Code:PA127770
Catalog # PIPA127770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human Clathrin light chain B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: Clathrin light chain B; clathrin, light chain (Lcb); clathrin, light chain B; clathrin, light polypeptide (Lcb); LCB
Gene Aliases: CLTB; LCB
UniProt ID: (Human) P09497
Entrez Gene ID: (Human) 1212
Molecular Function: membrane traffic protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.