Novus Biologicals
Manufacturer Code:NBP16894520UL
Catalog # NBP16894520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CLTB (clathrin light chain (Lcb)) The peptide sequence was selected from the C terminal of CLTB. Peptide sequence ADIIGYVASEEAFVKESKEETPGTEWEKVAQLCDFNPKSSKQCKDVSRLR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Clathrin light chain B; clathrin light chain B clathrin light chain (Lcb) clathrin light chain B clathrin light polypeptide (Lcb) Lcb; clathrin, light chain (Lcb); clathrin, light chain B; clathrin, light polypeptide (Lcb); Lcb
Gene Aliases: CLTB; LCB
UniProt ID: (Human) P09497
Entrez Gene ID: (Human) 1212
Molecular Function: membrane traffic protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.