Proteintech
Manufacturer Code:104551AP150UL
Catalog # 50-172-6321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CLTB Fusion Protein Ag0707 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Clathrin light chain B; Clathrin light chain B clathrin light chain (Lcb) CLTB LCB; clathrin, light chain (Lcb); clathrin, light chain B; clathrin, light polypeptide (Lcb); Lcb
Gene Aliases: 2310046E19Rik; CLTB; LCB
UniProt ID: (Human) P09497, (Rat) P08082, (Mouse) Q6IRU5
Entrez Gene ID: (Human) 1212, (Rat) 116561, (Mouse) 74325
Molecular Function:
membrane traffic protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.