Novus Biologicals
Manufacturer Code:NBP30323520UL
Catalog # NB095403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human CLS1 (NP_061968.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiolipin synthase (CMP-forming); cardiolipin synthase cardiolipin synthase 1 CLS dJ967N21.6; CLS; Protein GCD10 homolog
Gene Aliases: C20orf155; CLS; CLS1; CRLS1; dJ967N21.6; GCD10
UniProt ID: (Human) Q9UJA2
Entrez Gene ID: (Human) 54675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.