Invitrogen
Manufacturer Code:PA570444
Catalog # PIPA570444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human CLRN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Clarin-1; Clarin-1 Usher syndrome type-3 protein USH3A CLRN1; Usher syndrome 3A homolog; Usher syndrome type III A; Usher syndrome type-3 protein; Usher syndrome type-3 protein homolog
Gene Aliases: A130002D11Rik; CLRN1; RP61; USH3; USH3A
UniProt ID: (Human) P58418, (Mouse) Q8K445
Entrez Gene ID: (Horse) 100050427, (Dog) 611756, (Bovine) 538836, (Human) 7401, (Rabbit) 100342214, (Guinea pig) 100719258, (Mouse) 229320, (Rat) 261738
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.