Proteintech
Manufacturer Code:266301AP150UL
Catalog # 50-114-2752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CLRN1 Fusion Protein Ag24289 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: clarin 1 CLRN1 USH3 USH3A Usher syndrome type 3 protein; Clarin-1; Usher syndrome 3A homolog; Usher syndrome type-3 protein; Usher syndrome type-3 protein homolog
Gene Aliases: A130002D11Rik; CLRN1; RP61; USH3; USH3A
UniProt ID: (Human) P58418, (Mouse) Q8K445
Entrez Gene ID: (Human) 7401, (Mouse) 229320
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.