Invitrogen
Manufacturer Code:PA552722
Catalog # PIPA552722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CLPP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent Clp protease proteolytic subunit, mitochondrial; ATP-dependent protease ClpAP, proteolytic subunit, human; caseinolytic mitochondrial matrix peptidase proteolytic subunit; caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; caseinolytic protease, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic peptidase ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog; CLPP ClpP caseinolytic protease ATP-dependent proteolytic subunit homolog (E. coli) ATP-dependent proteolytic subunit E.coli homolog ATP-dependent protease ClpAP (E. coli) proteolytic subunit human ClpP (caseinolytic protease ATP-dependent proteolytic subunit E. coli) homolog ClpP caseinolytic peptidase ATP-dependent proteolytic subunit homolog (E. coli) ATP-dependent protease ClpAP proteolytic subunit human ClpP caseinolytic protease ATP-dependent proteolytic subunit homolog; Endopeptidase Clp; putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Gene Aliases: AU019820; CLPP; D17Wsu160e; DFNB81; PRLTS3
UniProt ID: (Human) Q16740, (Mouse) O88696
Entrez Gene ID: (Human) 8192, (Rat) 301117, (Mouse) 53895
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.