Novus Biologicals
Manufacturer Code:NBP131011
Catalog # NBP131011
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CLPP. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent Clp protease proteolytic subunit, mitochondrial; ATP-dependent protease ClpAP proteolytic subunit human ClpP (caseinolytic protease ATP-dependent proteolytic subunit E. coli)homolog ClpP caseinolytic peptidase ATP-dependent proteolytic subunit homolog (E.coli) ClpP caseinolytic protease ATP-dependent proteolytic subunit homolog ClpP caseinolytic protease ATP-dependent proteolytic subunit homolog (E.coli) EC 3.4.21.92 Endopeptidase Clp putative ATP-dependent Clp protease proteolytic subunit mitochondrial; ATP-dependent protease ClpAP, proteolytic subunit, human; ClpP caseinolytic peptidase ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog; Endopeptidase Clp; putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Gene Aliases: CLPP; DFNB81; PRLTS3
UniProt ID: (Human) Q16740
Entrez Gene ID: (Human) 8192
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.