Novus Biologicals
Manufacturer Code:NBP292342100UL
Catalog # NB026568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CLN8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C8orf61 ceroid-lipofuscinosis neuronal 8 (epilepsy progressive with mental retardation) EPMR; ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation); Protein CLN8
Gene Aliases: C8orf61; CLN8; EPMR
UniProt ID: (Human) Q9UBY8
Entrez Gene ID: (Human) 2055
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.