EMD Millipore
Manufacturer Code:MABN254825UG
Catalog # MABN254825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | N400/24 |
Immunogen | Recombinant fragments corresponding to 54 47 and 30 amino acids from non-helical regions from the N-terminal internal and C-terminal regions of human Ceroid-lipofuscinosis neuronal protein 6 (CLN6). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Stable for 1 year at 2-8°C from date of receipt. |
Mouse Monoclonal Antibody
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.