Novus Biologicals
Manufacturer Code:NBP15964620UL
Catalog # NBP15964620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CLN6(ceroid-lipofuscinosis neuronal 6 late infantile variant) The peptide sequence was selected from the middle region of CLN6. Peptide sequence LPRSITYVSIIIFIMGASIHLVGDSVNHRLLFSGYQHHLSVRENPIIKNL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ceroid-lipofuscinosis neuronal 6 late infantile; Ceroid-lipofuscinosis neuronal protein 6; ceroid-lipofuscinosis neuronal protein 6 ceroid-lipofuscinosis neuronal 6 late infantile variant FLJ20561 HsT18960 nclf Protein CLN6; Protein CLN6
Gene Aliases: CLN4A; CLN6; HsT18960; nclf
UniProt ID: (Human) Q9NWW5
Entrez Gene ID: (Human) 54982
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.