Invitrogen
Manufacturer Code:PA588837
Catalog # PIPA588837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 96-407 of human CLN5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceroid-lipofuscinosis neuronal protein 5; Ceroid-lipofuscinosis neuronal protein 5 homolog; Ceroid-lipofuscinosis neuronal protein 5 homolog, secreted form; Ceroid-lipofuscinosis neuronal protein 5, secreted form; Protein CLN5
Gene Aliases: A730075N08Rik; CLN5; NCL
UniProt ID: (Human) O75503, (Mouse) Q3UMW8
Entrez Gene ID: (Human) 1203, (Rat) 306128, (Mouse) 211286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.