Invitrogen
Manufacturer Code:PA5101484
Catalog # PIPA5101484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CLIP3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500005P14Rik AI844915 CAP-GLY domain containing linker protein 3 CAP-Gly domain-containing linker protein 3 clip-170 related protein 59 kDa CLIP-170-related 59 kDa protein CLIP3 CLIPR59 CLIPR-59 Cytoplasmic linker protein 170-related 59 kDa protein restin-like 1 RGD1306245 RSNL1; CAP-Gly domain-containing linker protein 3; clip-170 related protein 59 kDa; CLIP-170-related 59 kDa protein; Cytoplasmic linker protein 170-related 59 kDa protein; restin-like 1
Gene Aliases: 1500005P14Rik; AI844915; CLIP3; CLIPR-59; CLIPR59; RGD1306245; RSNL1
UniProt ID: (Human) Q96DZ5, (Mouse) B9EHT4
Entrez Gene ID: (Human) 25999, (Mouse) 76686, (Rat) 308493
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.