Novus Biologicals
Manufacturer Code:NBP2839750.1ML
Catalog # NB031904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human CLIP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAP-GLY domain containing linker protein 3 CAP-Gly domain-containing linker protein 3 CLIP-170-related 59 kDa protein CLIPR-59DKFZp586N1922 CLIPR59FLJ33413 Cytoplasmic linker protein 170-related 59 kDa protein restin-like 1 RSNL1; CAP-Gly domain-containing linker protein 3; CLIP-170-related 59 kDa protein; Cytoplasmic linker protein 170-related 59 kDa protein; restin-like 1
Gene Aliases: CLIP3; CLIPR-59; CLIPR59; RSNL1
UniProt ID: (Human) Q96DZ5
Entrez Gene ID: (Human) 25999
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.