Abnova
Manufacturer Code:PAB17644
Catalog # 89-119-308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human CLIP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CLIP3.
For Research Use Only
Protein Aliases: CAP-Gly domain-containing linker protein 3; CLIP-170-related 59 kDa protein; CLIPR-59; CLIPR59; Cytoplasmic linker protein 170-related 59 kDa protein; DKFZp586N1922; FLJ33413; restin-like 1; RSNL1
Gene Aliases: CLIP3; CLIPR-59; CLIPR59; RSNL1
UniProt ID: (Human) Q96DZ5
Entrez Gene ID: (Human) 25999
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.