Proteintech
Manufacturer Code:663431IG150UL
Catalog # 50-173-6774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B7F9 |
Immunogen | CLIC4 Fusion Protein Ag2942 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: chloride intracellular channel 4 (mitochondrial); chloride intracellular channel 4 like; Chloride intracellular channel protein 4; CLIC4 CLIC4L H1 huH1 MTCLIC p64H1; Intracellular chloride ion channel protein p64H1; mitochondrial chloride intracellular channel 4
Gene Aliases: CLIC4; CLIC4L; H1; huH1; MTCLIC; p64H1
UniProt ID: (Human) Q9Y696, (Rat) Q9Z0W7
Entrez Gene ID: (Human) 25932, (Pig) 100622787, (Rat) 83718
Molecular Function:
RNA binding protein
cytoskeletal protein
epimerase/racemase
isomerase
nucleic acid binding
oxidoreductase
reductase
signaling molecule
transferase
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.