Abnova
Manufacturer Code:PAB7536
Catalog # 89-133-344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human CLIC4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of CLIC4.
For Research Use Only
Protein Aliases: chloride intracellular channel 4 like; Chloride intracellular channel protein 4; CLIC4L; DKFZp566G223; FLJ38640; H1; huH1; Intracellular chloride ion channel protein p64H1; MTCLIC; p64H1
Gene Aliases: CLIC4; CLIC4L; H1; huH1; MTCLIC; p64H1
UniProt ID: (Human) Q9Y696
Entrez Gene ID: (Human) 25932
Molecular Function:
RNA binding protein
cytoskeletal protein
epimerase/racemase
isomerase
nucleic acid binding
oxidoreductase
reductase
signaling molecule
transferase
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.