Proteintech
Manufacturer Code:145451AP150UL
Catalog # 50-172-9533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CLIC1 Fusion Protein Ag6049 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chloride channel ABP; Chloride channel ABP CLIC1 G6 hRNCC NCC27; Chloride intracellular channel protein 1; chloride intracellular channel protein 1 {ECO:0000250|UniProtKB:O00299, ECO:0000312|EMBL:CAE83985.1}; clic1 {ECO:0000312|EMBL:CAE83985.1, ECO:0000312|RGD:1303043}; hRNCC; NCC27; Nuclear chloride ion channel 27; nuclear chloride ion channel protein; p64CLCP; Regulatory nuclear chloride ion channel protein; RNCC protein
Gene Aliases: Clcp; CLIC1; G6; NCC27
UniProt ID: (Human) O00299, (Mouse) Q9Z1Q5, (Rat) Q6MG61
Entrez Gene ID: (Human) 1192, (Mouse) 114584, (Rat) 406864
Molecular Function:
RNA binding protein
cytoskeletal protein
epimerase/racemase
isomerase
nucleic acid binding
oxidoreductase
reductase
signaling molecule
transferase
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.