Invitrogen
Manufacturer Code:PA542880
Catalog # PIPA542880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CLGN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2/6; Calmegin; Calnexin-T; MEG 1 antigen; testis tissue sperm-binding protein Li 79P
Gene Aliases: 4930459O04Rik; AI528775; CLGN; Cln; Meg1
UniProt ID: (Bovine) Q3SYT6, (Dog) E2RA18, (Human) O14967, (Mouse) P52194
Entrez Gene ID: (Rabbit) 100354203, (Bovine) 504256, (Dog) 476076, (Horse) 100063166, (Human) 1047, (Rat) 685504, (Guinea pig) 100720909, (Mouse) 12745
Molecular Function:
calcium-binding protein
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.