Invitrogen
Manufacturer Code:PA5100418
Catalog # PIPA5100418
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Calmegin corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4930459O04Rik A2/6 AI528775 Calmegin calnexin-T Clgn Cln MEG 1 antigen Meg1 testis tissue sperm-binding protein Li 79P; A2/6; Calmegin; Calnexin-T; MEG 1 antigen; testis tissue sperm-binding protein Li 79P
Gene Aliases: 4930459O04Rik; AI528775; CLGN; Cln; Meg1
UniProt ID: (Human) O14967, (Mouse) P52194
Entrez Gene ID: (Human) 1047, (Rat) 685504, (Mouse) 12745
Molecular Function: calcium-binding protein chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.