Proteintech
Manufacturer Code:181731AP150UL
Catalog # 50-173-2029
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CLEC4G Fusion Protein Ag12837 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 4 member G; C-type lectin domain family 4, member G; C-type lectin superfamily 4, member G; CLEC4G LP2698 LSECtin UNQ431; Liver and lymph node sinusoidal endothelial cell C-type lectin; LSECtin
Gene Aliases: CLEC4G; DTTR431; LP2698; LSECtin; UNQ431; UNQ431/PRO792
UniProt ID: (Human) Q6UXB4
Entrez Gene ID: (Human) 339390
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.