Invitrogen
Manufacturer Code:MA533341
Catalog # PIMA533341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | WEN43 |
Immunogen | Rat mincle-Fcgamma2b fusion protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type (calcium dependent carbohydrate-recognition domain) lectin superfamily member 9 C-type lectin domain family 4 member E C-type lectin domain family 4 member E C-type lectin superfamily member 9 CLEC4E CLECSF9 Macrophage-inducible C-type lectin MINCLE; C-type lectin domain family 4 member E; C-type lectin superfamily member 9; immunoreceptor; Macrophage-inducible C-type lectin; Mincle
Gene Aliases: Clec4e; Clecsf9; Mincle
UniProt ID: (Rat) Q67EQ1
Entrez Gene ID: (Rat) 450223
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.