Novus Biologicals
Manufacturer Code:NBP149311AF700
Catalog # NBP149311Z
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 16000 |
Immunogen | Recombinant human CLEC4E (41-219aa) purified from E. coli (NP_055173). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 9; C-type lectin domain family 4 member E; C-type lectin domain family 4, member E; C-type lectin superfamily member 9; CLECSF9 C-type (calcium dependent carbohydrate-recognition domain) lectin superfamilymember 9 C-type lectin domain family 4 member E C-type lectin domain family 4 member E Macrophage-inducible C-type lectin mincle MINCLEC-type lectin superfamily member 9; Macrophage-inducible C-type lectin; MINCLE
Gene Aliases: CLEC4E; CLECSF9; MINCLE; UNQ218/PRO244
UniProt ID: (Human) Q9ULY5
Entrez Gene ID: (Human) 26253
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.