Invitrogen
Manufacturer Code:PA5116924
Catalog # PIPA5116924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human CLEC4E |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 9; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 9; C-type lectin domain family 4 member E; C-type lectin domain family 4, member E; C-type lectin superfamily member 9; C-type lectin, superfamily member 9; C86253 CLEC4E CLECSF9 C-type (calcium dependent carbohydrate recognition domain) lectin superfamily member 9 C-type (calcium dependent carbohydrate-recognition domain) lectin superfamily member 9 C-type lectin domain family 4 member E C-type lectin domain family 4 member e C-type lectin superfamily member 9 C-type lectin superfamily member 9 immunoreceptor macrophage-inducible C-type lectin MINCLE UNQ218/PRO244; immunoreceptor; Macrophage-inducible C-type lectin; MINCLE
Gene Aliases: C86253; CLEC4E; CLECSF9; MINCLE; UNQ218/PRO244
UniProt ID: (Human) Q9ULY5, (Rat) Q67EQ1, (Mouse) Q9R0Q8
Entrez Gene ID: (Human) 26253, (Rat) 450223, (Mouse) 56619
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.