R&D Systems
Manufacturer Code:FAB2806G100UG
Catalog # FAB2806G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 413512 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human CLEC4D/CLECSF8 Gly52-Asn215 Accession # Q8WXI8 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member D; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin-like receptor 6; CD368; CD368 CLEC6 CLEC-6 CLECSF8 CLECSF8Mpcl C-type (calcium dependent carbohydrate-recognition domain) lectin superfamilymember 8 C-type lectin domain family 4 member D C-type lectin domain family 4 member D C-type lectin receptor C-type lectin superfamily member 8 C-type lectin-like receptor 6 macrophage C-type lectin MCL MGC40078 MPCL; CLEC-6; macrophage C-type lectin
Gene Aliases: CD368; CLEC-6; CLEC4D; CLEC6; CLECSF8; MCL; MPCL
UniProt ID: (Human) Q8WXI8
Entrez Gene ID: (Human) 338339
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.