R&D Systems
Manufacturer Code:MAB5170
Catalog # MAB5170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 805035 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human CLEC3B/Tetranectin Glu22-Val202 Accession # P05452 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 3 member B; C-type lectin domain family 3 member B C-type lectin domain family 3 member B DKFZp686H17246 Plasminogen kringle 4-binding protein TETN Tetranectin tetranectin (plasminogen binding protein) TNA TNAtetranectin (plasminogen-binding protein) TNtetranectin; C-type lectin domain family 3, member B; Plasminogen kringle 4-binding protein; Tetranectin; tetranectin (plasminogen-binding protein); TN
Gene Aliases: CLEC3B; TN; TNA
UniProt ID: (Human) P05452
Entrez Gene ID: (Human) 7123
Molecular Function:
extracellular matrix protein
extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.