Proteintech
Manufacturer Code:203111AP150UL
Catalog # 50-173-2822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CLEC3B Fusion Protein Ag14147 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 3 member B; C-type lectin domain family 3, member B; CLEC3B Tetranectin TN TNA; Plasminogen kringle 4-binding protein; Tetranectin; tetranectin (plasminogen binding protein); tetranectin (plasminogen-binding protein); TN
Gene Aliases: CLEC3B; TN; TNA
UniProt ID: (Human) P05452
Entrez Gene ID: (Human) 7123, (Mouse) 21922
Molecular Function:
extracellular matrix protein
extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.