Proteintech
Manufacturer Code:210131AP150UL
Catalog # 50-173-3202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CLEC18A Fusion Protein Ag15237 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 18 member A; C-type lectin domain family 18, member A; CLEC18A MRCL MRCL1 MRLP2; mannose receptor-like 1; Mannose receptor-like protein; Mannose receptor-like protein 2; mannose receptor-like protein 3
Gene Aliases: CLEC18A; MRCL; MRCL1; Mrlp; MRLP2; RGD1559899
UniProt ID: (Human) A5D8T8, (Mouse) Q7TSQ1
Entrez Gene ID: (Human) 348174, (Mouse) 353287, (Rat) 307851
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.