Proteintech
Manufacturer Code:144021AP150UL
Catalog # 50-172-9423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CLCNKA Fusion Protein Ag5647 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chloride channel K1; Chloride channel Ka; chloride channel Ka ClC K1 CLCK1 CLCNKA hClC Ka; Chloride channel protein ClC-Ka; chloride channel, kidney, A; chloride channel, voltage-sensitive Ka; ClC-K1; putative basolateral cTAL chloride channel ClC-Ka
Gene Aliases: C75963; CLC-K1; CLCK1; Clcnk1; CLCNKA; hClC-Ka
UniProt ID: (Human) P51800, (Mouse) Q9WUB7, (Rat) Q06393
Entrez Gene ID: (Human) 1187, (Mouse) 12733, (Rat) 79425
Molecular Function:
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.