Novus Biologicals
Manufacturer Code:NBP184452
Catalog # NBP184452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VVSRDSERLIGFAQRRELILAIKNARQRQEGIVSNSIMYFTEEPPELPANSPHPLKLRRILNLS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chloride channel 4; chloride channel 4 Chloride channel protein 4 Chloride transporter ClC-4 ClC-4 CLC4 ClC-4A H(+)/Cl(-) exchange transporter 4 MGC163150; Chloride channel protein 4; chloride channel, voltage-sensitive 4; Chloride transporter ClC-4; ClC-4; H(+)/Cl(-) exchange transporter 4
Gene Aliases: ClC-4; ClC-4A; CLC4; CLCN4
UniProt ID: (Human) P51793
Entrez Gene ID: (Human) 1183
Molecular Function: anion channel ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.