Novus Biologicals
Manufacturer Code:NBP259379H
Catalog # NBP259379H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S258-5 |
Immunogen | Synthetic peptide amino acids 98-115 (cytoplasmic N-terminus) of rat Clcn3 |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: chloride channel 3; chloride channel 3 Chloride channel protein 3 Chloride transporter ClC-3 ClC-3 CLC3 H(+)/Cl(-) exchange transporter 3; Chloride channel protein 3; chloride channel, voltage-sensitive 3; Chloride transporter ClC-3; ClC-3; H(+)/Cl(-) exchange transporter 3
Gene Aliases: ClC-3; CLC3; CLCN3
UniProt ID: (Human) P51790
Entrez Gene ID: (Human) 1182
Molecular Function:
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.