Invitrogen
Manufacturer Code:PA567906
Catalog # PIPA567906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human CLCN3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chloride channel 3; Chloride channel protein 3; chloride channel, voltage-sensitive 3; Chloride transporter ClC-3; ClC-3; H(+)/Cl(-) exchange transporter 3; protein kinase C-regulated chloride channel
Gene Aliases: ClC-3; CLC3; CLCN3
UniProt ID: (Human) P51790, (Rat) P51792, (Mouse) P51791
Entrez Gene ID: (Human) 1182, (Rat) 84360, (Mouse) 12725
Molecular Function:
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.